1-Benzyl-N-carbobenzoxy-L-glutamate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Cbz-L-glutamic Acid 1-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, Z-Glu-OBzl, 1-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester

B3989-5G B3989-25G
TCIAB3989-5GEA 279 DKK
TCIAB3989-5G TCIAB3989-25G
1-Benzyl-N-carbobenzoxy-L-glutamate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-N-carbobenzoxy-L-glutamate
Formel: C₂₀H₂₁NO₆
Molekylvægt: 371.39 g/mol
MDL Number: MFCD00077013
CAS-nummer: 3705-42-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -22.0 to -26.0 deg (C=1、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR