Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat
B3961-100MG
B3961-1G
TCIAB3961-100MGEA
1010
DKK
TCIAB3961-100MG
TCIAB3961-1G
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Formel:
C₁₆H₂₄BF₄Rh Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00075045 CAS-nummer: 35138-22-8 EINECS: 460-220-1 UN nummer: 1759 ADR: 8,III |
Specifikationer Testresultater
Appearance | Orange to Amber to Dark red powder to crystal |
Elemental analysis (Carbon) | 44.50 to 49.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...