1H-Benzotriazole-1-carboxaldehyde ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Formyl-1H-benzotriazole, 1H-Benzo[d][1,2,3]triazole-1-carbaldehyde
B3920-1G
B3920-5G
TCIAB3920-1GEA
723
DKK
TCIAB3920-1G
TCIAB3920-5G
1H-Benzotriazole-1-carboxaldehyde ≥95.0% (by GC)
1H-Benzotriazole-1-carboxaldehyde
Formel:
C₇H₅N₃O Molekylvægt: 147.14 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00239420 CAS-nummer: 72773-04-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...