2-Brom-4-nitrophenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-hydroxy-5-nitrobenzene

B3918-1G B3918-5G
TCIAB3918-1GEA 329 DKK
TCIAB3918-1G TCIAB3918-5G
2-Brom-4-nitrophenol ≥98.0% (by GC, titration analysis)
2-Brom-4-nitrophenol
Formel: C₆H₄BrNO₃
MDL Number: MFCD06656567
CAS-nummer: 5847-59-6
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR