2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Benzyl-2-(dimethylamino)-1-[4-(morpholino)phenyl]-1-butanone, 2-Benzyl-2-dimethylamino-1-(4-morpholinophenyl)-1-butanone

B3914-100G B3914-25G
TCIAB3914-100GEA 1160 DKK
TCIAB3914-100G TCIAB3914-25G
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone ≥98.0% (by HPLC, titration analysis)
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone
Formel: C₂₃H₃₀N₂O₂
MDL Number: MFCD00191775
CAS-nummer: 119313-12-1
EINECS: 404-360-3
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR