3,3'-Bithiophene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,3'-Bithienyl

B3906-1G B3906-5G
TCIAB3906-1GEA 706 DKK
TCIAB3906-1G TCIAB3906-5G
3,3'-Bithiophene ≥98.0% (by GC)
3,3'-Bithiophene
Formel: C₈H₆S₂
Molekylvægt: 166.26 g/mol
Kogepunkt: 180°C
MDL Number: MFCD00041214
CAS-nummer: 3172-56-3

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR