4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3886-1G B3886-100MG
TCIAB3886-1GEA 2300 DKK
TCIAB3886-1G TCIAB3886-100MG
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole ≥95.0% (by HPLC, total nitrogen)
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole
Formel: C₁₄H₆Br₂N₂S₃
MDL Number: MFCD16619295
CAS-nummer: 288071-87-4

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 245.0 to 249.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR