4-(Boc-amino)cyclohexanone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3856-1G B3856-5G
TCIAB3856-1GEA 165 DKK
TCIAB3856-1G TCIAB3856-5G
4-(Boc-amino)cyclohexanone ≥98.0% (by GC)
4-(Boc-amino)cyclohexanone
Formel: C₁₁H₁₉NO₃
MDL Number: MFCD00798168
CAS-nummer: 179321-49-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR