(R)-(+)-Butylene oxide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(R)-(+)-Ethylethylene Oxide, (R)-(+)-Ethyloxirane, (R)-(+)-1,2-Epoxybutane
B3832-25ML
B3832-5ML
TCIAB3832-25MLEA
2580
DKK
TCIAB3832-25ML
TCIAB3832-5ML
(R)-(+)-Butylene oxide ≥98.0% (by GC)
(R)-(+)-Butylene oxide
Formel:
C₄H₈O Kogepunkt: 63°C Flammepunkt: -22°C |
MDL Number:
MFCD02683444 CAS-nummer: 3760-95-0 UN nummer: 3022 ADR: 3,II |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | +9.0 to +11.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...