(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
HBPipU
B3816-5G
B3816-1G
TCIAB3816-5GEA
1150
DKK
TCIAB3816-5G
TCIAB3816-1G
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate ≥98.0% (by HPLC)
(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
Formel:
C₁₇H₂₄F₆N₅OP Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00191769 CAS-nummer: 190849-64-0 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 14.5 to 15.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...