1-Benzoylpiperidin-4-one ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B3778-5G
B3778-25G
TCIAB3778-5GEA
485
DKK
TCIAB3778-5G
TCIAB3778-25G
1-Benzoylpiperidin-4-one ≥97.0% (by GC)
1-Benzoylpiperidin-4-one
Formel:
C₁₂H₁₃NO₂ Kogepunkt: 160°C |
MDL Number:
MFCD00006189 CAS-nummer: 24686-78-0 EINECS: 246-407-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...