tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate, N-Boc-2,5-dihydropyrrole, N-BOC-3-Pyrroline
B3735-5G
B3735-25G
TCIAB3735-5GEA
583
DKK
TCIAB3735-5G
TCIAB3735-25G
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥98.0% (by GC)
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Formel:
C₉H₁₅NO₂ Kogepunkt: 75°C |
MDL Number:
MFCD01863512 CAS-nummer: 73286-70-1 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...