6-Bromo-1-indanone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3711-1G
B3711-5G
TCIAB3711-1GEA
345
DKK
TCIAB3711-1G
TCIAB3711-5G
6-Bromo-1-indanone ≥98.0% (by GC)
6-Bromo-1-indanone
Formel:
C₉H₇BrO Molekylvægt: 211.06 g/mol |
MDL Number:
MFCD02179286 CAS-nummer: 14548-39-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...