(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3700-1G B3700-5G
TCIAB3700-1GEA 165 DKK
TCIAB3700-1G TCIAB3700-5G
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0% (by GC, titration analysis)
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Formel: C₁₁H₁₉NO₄
MDL Number: MFCD00151904
CAS-nummer: 26250-84-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 122.0 to 126.0 deg-C
Specific rotation [a]20/D -44.0 to -48.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR