Benzyl-4-oxo-1-piperidinecarboxylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3643-25G B3643-5G
TCIAB3643-25GEA 1060 DKK
TCIAB3643-25G TCIAB3643-5G
Benzyl-4-oxo-1-piperidinecarboxylate ≥98.0% (by GC)
Benzyl-4-oxo-1-piperidinecarboxylate
Formel: C₁₃H₁₅NO₃
Kogepunkt: 170°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00673144
CAS-nummer: 19099-93-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR