N-tert-Butylmaleimide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3638-1G
B3638-5G
TCIAB3638-1GEA
321
DKK
TCIAB3638-1G
TCIAB3638-5G
N-tert-Butylmaleimide ≥98.0% (by GC)
N-tert-Butylmaleimide
Formel:
C₈H₁₁NO₂ Kogepunkt: 190°C Flammepunkt: 80°C |
MDL Number:
MFCD00191885 CAS-nummer: 4144-22-3 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...