Benzyl phenyl carbonate ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Carbonic Acid Benzyl Phenyl Ester
B3574-25G
B3574-5G
TCIAB3574-25GEA
1100
DKK
TCIAB3574-25G
TCIAB3574-5G
Benzyl phenyl carbonate ≥95.0% (by GC)
Benzyl phenyl carbonate
Formel:
C₁₄H₁₂O₃ Kogepunkt: 131°C |
MDL Number:
MFCD07784341 CAS-nummer: 28170-07-2 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...