sec-Butyl butyrate 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyric Acid sec-Butyl Ester

B3559-5ML B3559-25ML
TCIAB3559-5MLEA 460 DKK
TCIAB3559-5ML TCIAB3559-25ML
sec-Butyl butyrate 98,0 GC_ASSAY_METHOD
sec-Butyl butyrate
Formel: C₈H₁₆O₂
Molekylvægt: 144.21 g/mol
Kogepunkt: 152°C
CAS-nummer: 819-97-6
EINECS: 212-465-9
UN nummer: 3272
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR