N-Boc-azetidine-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid

B3540-1G B3540-5G
TCIAB3540-1GEA 517 DKK
TCIAB3540-1G TCIAB3540-5G
N-Boc-azetidine-3-carboxylic acid ≥98.0% (by GC, titration analysis)
N-Boc-azetidine-3-carboxylic acid
Formel: C₉H₁₅NO₄
Molekylvægt: 201.22 g/mol
MDL Number: MFCD01860897
CAS-nummer: 142253-55-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR