2-Bromo-3-hexylthiophene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3507-25G B3507-5G
TCIAB3507-25GEA 1620 DKK
TCIAB3507-25G TCIAB3507-5G
2-Bromo-3-hexylthiophene ≥98.0% (by GC)
2-Bromo-3-hexylthiophene
Formel: C₁₀H₁₅BrS
Kogepunkt: 75°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09907959
CAS-nummer: 69249-61-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR