1,3-Bis(3-hydroxy-3-pentyl)benzene ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3'-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha',alpha'-Tetraethyl-1,3-benzenedimethanol

B3436-25G B3436-5G
TCIAB3436-25GEA 3420 DKK
TCIAB3436-25G TCIAB3436-5G
1,3-Bis(3-hydroxy-3-pentyl)benzene ≥90.0% (by GC)
1,3-Bis(3-hydroxy-3-pentyl)benzene
Formel: C₁₆H₂₆O₂
Molekylvægt: 250.38 g/mol
MDL Number: MFCD14708166
CAS-nummer: 676465-94-4

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR