Bis(2-ethylhexyl)-3,3'-thiodipropionate ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
3,3'-Thiodipropionic Acid Bis(2-ethylhexyl) Ester
B3422-25G
B3422-500G
TCIAB3422-25GEA
132
DKK
TCIAB3422-25G
TCIAB3422-500G
Bis(2-ethylhexyl)-3,3'-thiodipropionate ≥96.0% (by GC)
Bis(2-ethylhexyl)-3,3'-thiodipropionate
Formel:
C₂₂H₄₂O₄S Kogepunkt: 180°C |
MDL Number:
MFCD01757653 CAS-nummer: 10526-15-5 EINECS: 234-083-1 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...