O-tert-Butyl-L-threonine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(2S,3R)-2-Amino-3-(tert-butoxy)butanoic acid, H-Thr(tBu)-OH
B3398-1G
B3398-5G
TCIAB3398-1GEA
132
DKK
TCIAB3398-1G
TCIAB3398-5G
O-tert-Butyl-L-threonine ≥98.0% (by titrimetric analysis)
O-tert-Butyl-L-threonine
Formel:
C₈H₁₇NO₃ Smeltepunkt: 260°C |
MDL Number:
MFCD00037766 CAS-nummer: 4378-13-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -42.0 to -46.0 deg (C=2, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...