Baclofen ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Amino-3-(4-chlorophenyl)butyric Acid, 4-Amino-3-(4-chlorophenyl)butanoic acid

B3343-25G B3343-5G B3343-1G
TCIAB3343-25GEA 1760 DKK
TCIAB3343-25G TCIAB3343-5G TCIAB3343-1G
Baclofen ≥98.0% (by titrimetric analysis)
Baclofen
Formel: C₁₀H₁₂ClNO₂
Molekylvægt: 213.66 g/mol
Smeltepunkt: 208°C
MDL Number: MFCD00055143
CAS-nummer: 1134-47-0
EINECS: 214-486-9
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,00937

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR