O-tert-Butyl-L-threonine-tert-butyl ester ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3340-25G B3340-5G
TCIAB3340-25GEA 1070 DKK
TCIAB3340-25G TCIAB3340-5G
O-tert-Butyl-L-threonine-tert-butyl ester ≥96.0% (by titrimetric analysis)
Formel: C₁₂H₂₅NO₃
Kogepunkt: 70°C
MDL Number: MFCD00077109
CAS-nummer: 5854-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=183e4757f01c521b2e557bdec13dd1b950582aed&vid=da108bdb099f13976afd4d576fb722e60749233e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +2.0 to +4.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR