(R)-1-Benzyl-3-pyrrolidinecarbonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-1-Benzyl-3-cyanopyrrolidine

B3299-1G B3299-5G
TCIAB3299-1GEA 1290 DKK
TCIAB3299-1G TCIAB3299-5G
(R)-1-Benzyl-3-pyrrolidinecarbonitrile ≥98.0% (by GC)
(R)-1-Benzyl-3-pyrrolidinecarbonitrile
Formel: C₁₂H₁₄N₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09951967
CAS-nummer: 157528-56-8

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -26.0 to -33.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR