1-Butyl-3-methylimidazoliummethansulphonat ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-n-Butyl-3-methylimidazolium methanesulphonate
B3231-5G
B3231-25G
TCIAB3231-5GEA
238
DKK
TCIAB3231-5G
TCIAB3231-25G
1-Butyl-3-methylimidazoliummethansulphonat ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazoliummethansulphonat
Formel:
C₉H₁₈N₂O₃S |
MDL Number:
MFCD06798173 CAS-nummer: 342789-81-5 UN nummer: 3261 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...