N-(tert-Butoxycarbonyl)-L-2-phenylglycine ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Boc-Phg-OH, BOC-L-alpha-phenylglycine, Boc-D-Phg-OH, N-Boc-L-2-phenylglycine

B3211-25G B3211-5G
TCIAB3211-25GEA 583 DKK
TCIAB3211-25G TCIAB3211-5G
N-(tert-Butoxycarbonyl)-L-2-phenylglycine ≥98.0% (by HPLC)
N-(tert-Butoxycarbonyl)-L-2-phenylglycine
Formel: C₁₃H₁₇NO₄
MDL Number: MFCD00065588
CAS-nummer: 2900-27-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 91.0 to 95.0 deg-C
Specific rotation [a]20/D +137.0 to +147.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR