1-Benzylpiperidine-4-carbaldehyde ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Benzylpiperidine-4-carboxaldehyde, 1-Benzyl-4-piperidinecarboxaldehyde

B3190-25G B3190-5G
TCIAB3190-25GEA 1160 DKK
TCIAB3190-25G TCIAB3190-5G
1-Benzylpiperidine-4-carbaldehyde ≥96.0% (by GC, titration analysis)
1-Benzylpiperidine-4-carbaldehyde
Formel: C₁₃H₁₇NO
Kogepunkt: 128°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02258902
CAS-nummer: 22065-85-6
EINECS: 244-757-7

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR