5-Bromo-2-adamantanone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5-Bromoadamantan-2-one
B3154-1G
B3154-5G
TCIAB3154-1GEA
723
DKK
TCIAB3154-1G
TCIAB3154-5G
5-Bromo-2-adamantanone ≥98.0% (by GC)
5-Bromo-2-adamantanone
Formel:
C₁₀H₁₃BrO |
MDL Number:
MFCD01321097 CAS-nummer: 20098-20-8 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...