4-(tert-Butoxycarbonylamino)-1-butanol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate
B2868-25G
B2868-5G
TCIAB2868-25GEA
2070
DKK
TCIAB2868-25G
TCIAB2868-5G
4-(tert-Butoxycarbonylamino)-1-butanol ≥96.0% (by GC)
4-(tert-Butoxycarbonylamino)-1-butanol
Formel:
C₉H₁₉NO₃ Molekylvægt: 189.26 g/mol |
MDL Number:
MFCD01861334 CAS-nummer: 75178-87-9 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...