4,4'-Biphenyldicarboxaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: [1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl

B2854-1G B2854-5G
TCIAB2854-1GEA 403 DKK
TCIAB2854-1G TCIAB2854-5G
4,4'-Biphenyldicarboxaldehyde ≥98.0% (by GC)
4,4'-Biphenyldicarboxaldehyde
Formel: C₁₄H₁₀O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00016714
CAS-nummer: 66-98-8

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR