(2-Benzimidazolyl)acetonitrile ≥96.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B2821-25G
B2821-5G
TCIAB2821-25GEA
1040
DKK
TCIAB2821-25G
TCIAB2821-5G
(2-Benzimidazolyl)acetonitrile ≥96.0% (by GC, titration analysis)
(2-Benzimidazolyl)acetonitrile
Formel:
C₉H₇N₃ Smeltepunkt: 212°C |
MDL Number:
MFCD00005601 CAS-nummer: 4414-88-4 EINECS: 224-574-9 |
Specifikationer Testresultater
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...