18-Bromo-1-octadecene ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 17-Octadecenyl Bromide

B2816-1G B2816-5G
TCIAB2816-1GEA 1370 DKK
TCIAB2816-1G TCIAB2816-5G
18-Bromo-1-octadecene ≥93.0% (by GC)
18-Bromo-1-octadecene
Formel: C₁₈H₃₅Br
Opbevaringstemperatur: Køleskab
CAS-nummer: 172656-11-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR