Benzyl dimethylphosphonoacetate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate

B2815-1G B2815-5G
TCIAB2815-1GEA 312 DKK
TCIAB2815-1G TCIAB2815-5G
Benzyl dimethylphosphonoacetate ≥97.0% (by GC)
Benzyl dimethylphosphonoacetate
Formel: C₁₁H₁₅O₅P
MDL Number: MFCD01872507
CAS-nummer: 57443-18-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR