Benzyl-4-bromobutyl ether ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Benzyl 4-bromobutyl ether, (4-Bromobutoxymethyl)benzene, ((4-Bromobutoxy)methyl)benzene, 4-Benzyloxybutyl Bromide

B2802-25G B2802-5G
TCIAB2802-25GEA 4320 DKK
TCIAB2802-25G TCIAB2802-5G
Benzyl-4-bromobutyl ether ≥96.0% (by GC)
Benzyl-4-bromobutyl ether
Formel: C₁₁H₁₅BrO
Molekylvægt: 243.14 g/mol
Kogepunkt: 115°C
MDL Number: MFCD00155128
CAS-nummer: 60789-54-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR