9,10-Bis(diethylphosphonomethyl)anthracene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
[Anthracene-9,10-diylbis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [Anthracene-9,10-diylbis(methylene)]bisphosphonate, Tetraethyl (anthracene-9,10-diylbis(methylene))bis(phosphonate)
B2801-1G
B2801-5G
TCIAB2801-1GEA
517
DKK
TCIAB2801-1G
TCIAB2801-5G
9,10-Bis(diethylphosphonomethyl)anthracene ≥98.0% (by GC)
9,10-Bis(diethylphosphonomethyl)anthracene
Formel:
C₂₄H₃₂O₆P₂ Molekylvægt: 478.46 g/mol |
MDL Number:
MFCD08276315 CAS-nummer: 60974-92-7 |
Specifikationer Testresultater
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...