4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B2773-1G B2773-5G
TCIAB2773-1GEA 485 DKK
TCIAB2773-1G TCIAB2773-5G
4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)
4-Bromoindole-3-carboxyaldehyde
Formel: C₉H₆BrNO
Molekylvægt: 224.06 g/mol
MDL Number: MFCD05864695
CAS-nummer: 98600-34-1

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %
Melting point 177.0 and le 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR