Bis(2,2,2-trifluoroethyl)phosphite ≥94.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Phosphorous Acid Bis(2,2,2-trifluoroethyl) Ester, Bis(2,2,2-trifluoroethyl) Phosphonate, Phosphonic Acid Bis(2,2,2-trifluoroethyl) Ester
B2627-25G
B2627-5G
TCIAB2627-25GEA
1040
DKK
TCIAB2627-25G
TCIAB2627-5G
Bis(2,2,2-trifluoroethyl)phosphite ≥94.0% (by GC)
Bis(2,2,2-trifluoroethyl)phosphite
Formel:
C₄H₅F₆O₃P Molekylvægt: 246.05 g/mol Kogepunkt: 44°C Flammepunkt: 76°C |
MDL Number:
MFCD00063314 CAS-nummer: 92466-70-1 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...