N,N-Bis(2-isopropoxyethyl)amine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B2356-25G B2356-5G
TCIAB2356-25GEA 1780 DKK
TCIAB2356-25G TCIAB2356-5G
N,N-Bis(2-isopropoxyethyl)amine ≥97.0% (by GC, titration analysis)
N,N-Bis(2-isopropoxyethyl)amine
Formel: C₁₀H₂₃NO₂
MDL Number: MFCD06797066
CAS-nummer: 1119451-05-6
UN nummer: 2735
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR