1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate) ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B2280-100MG B2280-500MG
TCIAB2280-100MGEA 1330 DKK
TCIAB2280-100MG TCIAB2280-500MG
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate) ≥98.0% (by HPLC)
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate)
Formel: C₃₄H₂₈F₁₂N₄P₂
Smeltepunkt: 154°C
MDL Number: MFCD06797055
CAS-nummer: 134815-78-4

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR