4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-(4-Biphenylyl)-N,N-bis[4-[di(3,5-xylyl)amino]phenyl]amine
B2269-1G
B2269-100MG
TCIAB2269-1GEA
9250
DKK
TCIAB2269-1G
TCIAB2269-100MG
4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine ≥98.0% (by HPLC)
4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine
Formel:
C₅₆H₅₃N₃ Molekylvægt: 768.06 g/mol |
MDL Number:
MFCD03844780 CAS-nummer: 249609-49-2 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...