3-(Boc-amino)propylbromide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester
B2234-1G
B2234-5G
TCIAB2234-1GEA
362
DKK
TCIAB2234-1G
TCIAB2234-5G
3-(Boc-amino)propylbromide ≥98.0% (by titrimetric analysis)
3-(Boc-amino)propylbromide
Formel:
C8H16BrNO2 Molekylvægt: 238.1221 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD02683429 CAS-nummer: 83948-53-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 36.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...