(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(-)-2,6-Bis[(4S)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline), 2,6-Bis((S)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (S)-(i-Pr)-pybox
B2218-5G
B2218-500MG
TCIAB2218-5GEA
3300
DKK
TCIAB2218-5G
TCIAB2218-500MG
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0% (by GC)
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine
Formel:
C₁₇H₂₃N₃O₂ |
MDL Number:
MFCD00191815 CAS-nummer: 118949-61-4 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 151.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...