Bis[ɑ,ɑ-bis(trifluoromethyl)benzenemethanolato]diphenylsulphur ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Martin Sulphurane

B2200-1G B2200-5G
TCIAB2200-1GEA 575 DKK
TCIAB2200-1G TCIAB2200-5G
Bis[ɑ,ɑ-bis(trifluoromethyl)benzenemethanolato]diphenylsulphur ≥96.0% (by titrimetric analysis)
Bis[alpha,alpha-bis(trifluoromethyl)benzenemethanolato]diphenylsulphur
Formel: C₃₀H₂₀F₁₂O₂S
Molekylvægt: 672.53 g/mol
Smeltepunkt: 111°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00010662
CAS-nummer: 32133-82-7
UN nummer: 3261
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR