1-[Bis(2-hydroxyethyl)amino]-2-propanol ≥93.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: N,N-Bis(2-hydroxyethyl)isopropanolamine
B2180-25G B2180-5G
TCIAB2180-25GEA 2620 DKK
TCIAB2180-25G TCIAB2180-5G
1-[Bis(2-hydroxyethyl)amino]-2-propanol ≥93.0% (by GC)
1-[Bis(2-hydroxyethyl)amino]-2-propanol
Formel: C₇H₁₇NO₃
Molekylvægt: 163.22 g/mol
Kogepunkt: 145°C
Flammepunkt: 113°C
MDL Number: MFCD00009725
CAS-nummer: 6712-98-7
EINECS: 229-764-5
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 93.0 %