4,4'-Bis(bromomethyl)biphenyl ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4,4-Bis(bromomethyl)biphenyl
B1921-1G
B1921-5G
B1921-25G
TCIAB1921-1GEA
345
DKK
TCIAB1921-1G
TCIAB1921-5G
TCIAB1921-25G
4,4'-Bis(bromomethyl)biphenyl ≥95.0% (by GC)
4,4'-Bis(bromomethyl)biphenyl
Formel:
C₁₄H₁₂Br₂ |
MDL Number:
MFCD00094419 CAS-nummer: 20248-86-6 UN nummer: 1759 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...