Ethyl-7-bromoheptanoate ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
7-Bromoheptanoic Acid Ethyl Ester, Ethyl 7-bromoheptanoate
B1880-25G
B1880-5G
TCIAB1880-25GEA
4510
DKK
TCIAB1880-25G
TCIAB1880-5G
Ethyl-7-bromoheptanoate ≥97.0% (by GC)
Ethyl-7-bromoheptanoate
Formel:
C₉H₁₇BrO₂ Kogepunkt: 145°C |
MDL Number:
MFCD00037103 CAS-nummer: 29823-18-5 EINECS: 249-883-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...