1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Benzyl N-Boc-L-glutamate, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, N-Boc-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl

B1864-5G B1864-25G
TCIAB1864-5GEA 370 DKK
TCIAB1864-5G TCIAB1864-25G
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate ≥98.0% (by titrimetric analysis)
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate
Formel: C₁₇H₂₃NO₆
Molekylvægt: 337.37 g/mol
MDL Number: MFCD00065568
CAS-nummer: 30924-93-7

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C
Specific rotation [a]20/D -28.0 to -31.0 deg (C=0.7, MeOH)
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR