3-(2-Methoxyethoxy)propyl bromide ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-3-(2-methoxyethoxy)propane, Ethylene Glycol 3-Bromopropyl Methyl Ether
B1842-1G
B1842-5G
TCIAB1842-1GEA
624
DKK
TCIAB1842-1G
TCIAB1842-5G
3-(2-Methoxyethoxy)propyl bromide ≥95.0% (by GC)
3-(2-Methoxyethoxy)propyl bromide
Formel:
C₆H₁₃BrO₂ Molekylvægt: 197.07 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01321130 CAS-nummer: 59551-75-6 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...